Learn More
Medchemexpress LLC PrP (106-126) | 148439-49-0 | 99.6% | 1912.30 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP030510MG
Encompass_Preferred
PrP (106-126) is a peptide corresponding to the prion protein (PrP) amyloidogenic region. Its biochemical properties are similar to the infectious form of prion protein. This product is for research use only.
  • Corresponds to the prion protein (PrP) amyloidogenic region
  • Biochemical properties resemble the infectious form of prion protein
  • Induces mTOR phosphorylation in N2a cells
  • Increases ROS production
  • Interacts selectively with porcine brain endothelial cells
  • Reduces trans-endothelial electrical resistance
Catalog No. 50-004-24881