Learn More
ATCC pBAD33 phagemid; in E. coli
Supplier:  ATCC 87402
Encompass
pBAD33 is a low copy number expression vector regulated by the arabinose operon. It is useful when reduced gene expression is desired.
Catalog No. 50-238-1516