Learn More
Selleck Chemical LLC PRT-060318 2HCl
SDP
Supplier:  Selleck Chemical LLC S773825MG
Encompass_Preferred
PRT-060318 (PRT318) is a novel selective inhibitor of the Syk tyrosine kinase with an IC50 of 4 nM as an approach to HIT treatment
Catalog No. 50-000-01067