Learn More
Medchemexpress LLC PRT062607 | 1370261-96-3 | C19H23N9O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1532250MG
Encompass_Preferred
PRT062607 is an orally active ATP-competitive Syk inhibitor with an IC50 of 1 nM, demonstrating at least 80-fold selectivity over other kinases. It blocks B cell antigen receptor-mediated activation, Fcε receptor 1-mediated basophil degranulation and microglial phagocytosis, and induces caspase-dependent apoptosis and microglial death. It inhibits tumor growth and peripheral nerve injury-induced mechanical allodynia, and prevents neuronal loss.
  • Blocks B cell antigen receptor-mediated activation, Fcε receptor 1-mediated basophil degranulation, and microglial phagocytosis.
  • Induces caspase-dependent apoptosis and microglial death.
  • Inhibits tumor growth and peripheral nerve injury-induced mechanical allodynia.
  • Prevents neuronal loss.
  • Used in research for rheumatoid arthritis, chronic lymphocytic leukemia, non-Hodgkin's lymphoma, neurodegenerative diseases, and neuropathic pain.
Catalog No. 50-003-72640