Learn More
Apexbio Technology LLC PRT062607 Hydrochloride,5mg CAS# 1370261-97-4
SDP
Supplier:  Apexbio Technology LLC A37365
Encompass_Preferred
PRT062607 Hydrochloride is a highly selective and novel SYK inhibitor with IC50 value of 1 nM. Spleen tyrosine kinase (SYK) is a cytoplasmic tyrosine kinase and primarily expressed in hematopoietic cells including B-cells.PRT062607 Hydrochloride is an orally bioavailable SYK inhibitor and its affinity for SYK is at least 80-fold greater than other kinases. In NHL cell lines, PRT062607 Hydrochloride inhibits SYK activation and induces caspase dependent apoptosis. Also, PRT062607 Hydrochloride inhibited SYK and caused apoptosis of the tumor B-cell line [1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0164