Learn More
Enzo Life Sciences Proteasome 20S beta5 subunit (human) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW88950025
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 194-207 of human proteasome 20S (β5 subunit). UniProt: P28074. Source: Purified from rabbit serum. Species Reactivity: Human. Specificity: Recognizes human proteasome 20S β5 subunit. Applications: IHC, WB. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. NC0263076