Learn More
TARGETMOL CHEMICALS INC PRUCALOPRIDE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2542100MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Prucalopride (R-93877) is a selective high-affinity 5-HT4A/4B receptor agonist (Ki 2.5/8 nM). It has >290-fold selectivity for 5-HT4A/4B receptor than other 5-HT receptor subtypes. purity: 100%
Catalog No. 50-257-7095