Learn More
Apexbio Technology LLC Prucalopride, 10mg. Cas: 179474-81-8 MFCD: MFCD09837787. 5-HT4A and 5-HT4B receptor agonist
SDP
Supplier:  Apexbio Technology LLC B225310
Encompass_Preferred
Prucalopride is a selective, high affinity 5-HT receptor agonist for 5-HT4A and 5-HT4B receptor with Ki of 2.5 nM and 8 nM, respectively, exhibits >290-fold selectivity against other 5-HT receptor subtypes. Phase 3. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5334
 Restricted Item