Learn More
Medchemexpress LLC Prucalopride-13C,d3 | 2140306-00-7 | 99.0% | C1713CH23D3ClN3O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14151S10MG
Encompass_Preferred
Prucalopride-13C,d3 is a stable isotope-labeled analogue of prucalopride that incorporates 13C and deuterium labels. It is provided as a solid research reagent for use as an internal standard, tracer in metabolic and pharmacokinetic studies, and for analytical method development. For research use only; not for human or clinical use.
  • Stable isotope labeled for tracer and internal standard applications.
  • High chemical purity suitable for quantitative analysis.
  • Provided in small milligram quantities for method development and PK studies.
  • Supplied as a powder with recommended low-temperature storage.
  • Compatible with LC-MS/MS workflows for quantitation and metabolic profiling.
Catalog No. 50-002-58890