Learn More
Medchemexpress LLC Prucalopride (succinate) | 179474-85-2 | MFCD00948731 | 100.0% | 485.96 g·mol⁻¹ | C22H32ClN3O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY126945MG
Encompass_Preferred
Prucalopride (succinate) is the succinate salt of prucalopride, a selective, high-affinity 5-HT4 receptor agonist supplied as a research-grade analytical standard. It is intended for use in studies of gastrointestinal motility, 5-HT4 pharmacology, and related in vitro and in vivo research, and is characterized with accompanying COA and SDS documentation.
  • Selective 5-HT4 receptor agonist suitable for pharmacological studies.
  • High purity analytical standard for reproducible results.
  • Available in small solid quantities for analytical and preclinical work.
  • Supports in vitro and in vivo studies of intestinal motility and related signaling pathways.
  • Documented with COA and SDS for traceability and quality control.
Catalog No. 50-002-64469