Learn More
Medchemexpress LLC Prusogliptin | 1186426-66-3 | 99.9% | 324.39 | C16H25FN4O2 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY1252810MM 1ML
Encompass_Preferred
DBPR108 (Prusogliptin) is a potent, selective dipeptidyl peptidase-4 (DPP-4) inhibitor supplied for research use. It exhibits an IC50 of 15 nM against DPP-4, shows no inhibition of DPP8 or DPP9, and has been investigated as an oral candidate for type 2 diabetes. The compound is provided as a solid or as a 10 mM solution in DMSO, with supporting analytical documentation.
Catalog No. 50-002-35080