Learn More
Medchemexpress LLC Prussian blue soluble | 12240-15-2 | MFCD00135664 | 306.89 | C6Fe2KN6 | 1 G
SDP
Supplier:  Medchemexpress LLC HY106594B1G
Encompass_Preferred
Prussian blue soluble is a potassium ferric ferrocyanide inorganic pigment (Pigment Blue 27) supplied as a soluble blue powder for laboratory research. It is commonly used as a histological stain to detect ferric iron and as an adsorbent in studies involving cesium or thallium ions. The material is provided in small research pack sizes for bench-top use.
Catalog No. 50-002-40061