Learn More
Medchemexpress LLC Prussian blue soluble | 12240-15-2 | MFCD00135664 | 306.89 g/mol | C6Fe2KN6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY106594B100MG
Encompass_Preferred
Prussian blue soluble is a research-grade inorganic pigment and adsorbent supplied as a blue solid for laboratory use. It is used as an adsorbent antidote for cesium and thallium poisoning, is investigated for anticancer and antibacterial effects, and can function as a contrast agent in photoacoustic and magnetic resonance imaging. Supplied in multiple research pack sizes; for research use only.
Catalog No. 50-002-40995