Learn More
Associates of Cape Cod Inc PRYOTELL-T LAL 5ML 5PK
SDP
Supplier:  Associates of Cape Cod Inc T00515
Encompass
PYROTELL-T TURBIDIMETRIC LAL, SENSITIVE TO 0.001 EU/mL WHEN USED WITH ACC TUBE READERS, 5 mL/VIAL, 5 VIALS/PACK
Catalog No. NC0233934