Learn More
AdipoGen Bortezomib, 179324-69-7, MFCD09056737, 5 mg
SDP
Supplier:  AdipoGen AGCR13602M005
Encompass_Preferred
MF: C19H25BN4O4, MW: 384.2, ≥97%, Synonyms: PS-341; LDP-341; MLM-341; MG-341; NSC-681239; Proteasome Inhibitor. Appearance: White solid. Highly potent, selective and reversible cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Anticancer compound. Inhibits proliferation and migration of several tumor cell lines with nanomolar potency. Apoptosis inducer.
Catalog No. 50-101-6263