Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARTICLE MEASURING SYSTEMS INC PS R 150W 19V UNIVERSAL

Supplier:  PARTICLE MEASURING SYSTEMS INC 1000024952

Encompass

  • Universal power supply for airborne particle counters
  • Provides reliable external power
  • Ensures uninterrupted operation of cleanroom monitoring equipment
  • Facilitates simultaneous charging of onboard batteries
  • Robust design for scientific environments
  • Supports 150 W output power
  • Operates at 19 V output voltage
  • Compatible with Lasair Pro and Lasair III models

Catalog No. NC3397501


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.