Learn More
Medchemexpress LLC Ps10 | 1564265-82-2 | 98.5% | 10 MG

Supplier: Medchemexpress LLC HY12174410MG
PS10 is a small-molecule, ATP-competitive pan-pyruvate dehydrogenase kinase (PDK) inhibitor used in research to probe metabolic pathways and cardiac metabolism. It has been reported to improve glucose tolerance and stimulate myocardial carbohydrate oxidation in diet-induced obesity models.
- ATP-competitive pan-PDK inhibitor.
- Inhibits PDK isoforms with IC50s: PDK2 0.8 μM, PDK4 0.76 μM, PDK1 2.1 μM, PDK3 21.3 μM.
- High reported purity of 98.5%.
- Molecular weight 323.32 g·mol⁻1, formula C14H13NO6S.
- Supplied as a solid 10 mg and as 10 mM solution in DMSO for convenience.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.