Learn More
Medchemexpress LLC PS372424 (hydrochloride) | 1596362-29-6 | ≥98.0% | 625.20 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111149A
Encompass_Preferred
PS372424 hydrochloride, a three amino-acid fragment of CXCL10, is a specific human CXCR3 agonist with anti-inflammatory activity. It prevents human T-cell migration in a humanized model of arthritic inflammation.
  • Specific human CXCR3 agonist with anti-inflammatory activity.
  • Prevents human T-cell migration in a humanized model of arthritic inflammation.
Catalog No. 50-003-45770