Learn More
Sino Biological Peptide Substrates: PS7-CTD Peptide
Supplier:  Sino Biological R065801
Encompass_Preferred
The PS7-CTD peptide sequence (YSPTS PpSYSP TSPpS) is derived from C-terminal repeat domain of human RNA polymerase II.
Catalog No. 50-256-8009