Learn More
Genscript Corporation PSA 8A12 mAb Mouse
Supplier:  Genscript Corporation V07301100
Encompass
Prostate specific antigen (PSA) is a serine protease produced by the prostate gland In serum it exists in different forms cPSA mainly including PSA-ACT (alpha-1-antichymotrypsin) and PSA-A2M (alpha-2-macroglobulin) fPSA (free PSA) etc The blood level of PSA is increased in men with prostate cancer and it is considered as an indicator for prostate cancer tumor recurrence and response to therapy
Catalog No. 50-000-58294
Only null left
Add to Cart
Request a Quote