Learn More
Genetex Inc PSA Monoclonal Antibody [MSVA-603R] HistoMAX™ (GTX04481) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04481
Encompass_Preferred
The PSA antibody [MSVA-603R] HistoMAX™ (GTX04481) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human kidney tissue using GTX04481 PSA antibody [MSVA-603R] HistoMAX. Absence of PSA immunostaining.
Catalog No. NC3667791