Learn More
Genscript Corporation PSA Antibody 8A9B8 mAb Mou
Supplier:  Genscript Corporation A016645
Encompass
PSA monoclonal antibodies (5B10D4 8A9B8 1G9G8 4A5D10 and 1A7D4) recognize total PSA freePSA and ACT- PSA Complex (Prostate Specific Antigen / a-1 Antichymotrypsin Complex)
Catalog No. 50-000-57074