Learn More
Medchemexpress LLC PSAP/Prosaposin Mou 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77159100UG
Encompass_Preferred
PSAP/Prosaposin protein is a myelin trophic factor and neurotrophic factor that acts through GPR37 and GPR37L1 receptors This triggers ligand-mediated internalization initiating ERK phosphorylation signaling PSAP/Prosaposin Protein Mouse (HEK293 His) is the recombinant mouse-derived PSAP/Prosaposin protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86495