Learn More
Medchemexpress LLC PSB 0777 ammonium | >=97.0% | 500.55 | C18H24N6O7S2 | 5 MG

Supplier: Medchemexpress LLC HY136233A5MG
PSB 0777 ammonium is a potent, selective adenosine A2A receptor full agonist used as a research reagent to study A2A-mediated signaling and pharmacology. It is supplied in small quantities for laboratory use and has reported nanomolar activity in receptor binding assays.
- Potent and selective adenosine A2A receptor full agonist.
- Nanomolar binding activity to A2A receptors.
- Suitable for in vitro and in vivo research applications.
- Supplied in small quantity packages, typically 1 mg and 5 mg.
- Purity ≥97% (HPLC) as reported by independent suppliers.
- Chemical formula C18H24N6O7S2; molecular weight 500.55.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.