Learn More
Abcam PSB 11 hydrochloride, competitive human A3 receptor antagonist, 10MG
Supplier:  Abcam AB12099510MG
Encompass_Preferred
MW 331.8 Da, Purity >99%. Potent, selective, competitive human A3 receptor antagonist (Ki values are 2.3 nM and 10 μM for human and rat A3 receptors, respectively), Antagonises human A1 and A2A receptors. Shows inverse agonist effects in the [35S]GTPγS binding assay. Shows anti-inflammatory effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7050