Learn More
Medchemexpress LLC Psb-12379 disodium | 1802226-78-3 | 100.0% | 515.35 | C18H23N5O9P2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10074725MG
Encompass_Preferred
PSB-12379 is a nucleotide analogue and potent inhibitor of ecto-5'-nucleotidase (CD73), showing low-nanomolar activity in human and rat assays. The compound is supplied with high HPLC purity and is provided in solid and solution formats for research use. Recommended storage conditions and handling guidance are provided for both powder and solvent preparations.
Catalog No. 50-002-12097