Learn More
Medchemexpress LLC Psb-603 | 1092351-10-4 | 99.4% | 529.01 | C24H25ClN6O4S | 10 MG

Supplier: Medchemexpress LLC HY10316610MG
PSB-603 is a potent and selective adenosine A2B receptor antagonist used in pharmacological research to probe A2B-mediated signaling. It exhibits subnanomolar affinity at human A2B receptors and is supplied as a solid research reagent.
- High potency at A2B receptor (Ki = 0.553 nM).
- Minimal affinity for A1, A2A, and A3 receptors up to 10 μM.
- Chemical formula C24H25ClN6O4S; molecular weight 529.01.
- High purity (99.38%); solid, off-white to light brown.
- Suitable for in vitro and in vivo pharmacological studies.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.