Learn More
Medchemexpress LLC PSEM 89S TFA | 1336913-03-1 | 99.9% | 404.38 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY112217A50MG
Encompass_Preferred
PSEM 89S TFA is a selective and brain penetrant agonist for the resulting ion channels. It is orthogonally selective for Q79G and L141F, respectively. This product is for research use only.
  • Selective and brain penetrant agonist for ion channels.
  • Orthogonally selective for Q79G and L141F.
Catalog No. 50-004-38354