Learn More
American Radiolabeled Chemicals Inc PSER L-DIOL-1-14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 363550UC
Encompass_Preferred
Phosphatidylserine, L-a-dioleoyl-oleoyl-1-14C, 50uCi in ethanol:toluene (1:1), 0.1 mCi/ml, 50-60 mCi/mmol, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-980
Add to Cart
Request a Quote