Learn More
Medchemexpress LLC Pseudobaptigenin | 90-29-9 | 282.25 | 5 MG

Supplier: Medchemexpress LLC HYN106165MG
Pseudobaptigenin is a flavonoid that exhibits very good anticataract activity. It demonstrates strong binding affinity for inhibiting glycation against γ-crystallin protein and possesses good ADMET (Absorption, Distribution, Metabolism, Excretion, and Toxicity) properties. It is intended for research use only.
- Very good anticataract activity
- Good binding affinity for the inhibition of glycation against γ-crystallin protein
- Good ADMET properties
- Purity: 98.41%
- Appearance: Solid
- Color: Light yellow to light brown
- Structure classification: Flavonoids, Flavones
- Initial source: Plants (Leguminosae, Pterocarpus marsupium Roxb.)
- Solubility (in vitro): DMSO: 35.71 mg/mL (126.52 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.