Learn More
Medchemexpress LLC Pseudoginsenoside RT1 | 98474-74-9 | 99.5% | 927.08 | C47H74O18 | 10 MG

Supplier: Medchemexpress LLC HYN220110MG
Pseudoginsenoside RT1 is a natural saponin isolated from the fruit of Randia siamensis and is supplied as a research-grade reference standard. It exhibits reported acute ichthyotoxic activity and is intended for analytical and laboratory use only.
- Natural saponin isolated from Randia siamensis.
- Reported acute ichthyotoxic activity.
- High purity (≈99.5%).
- Molecular formula C47H74O18; molecular weight 927.08.
- CAS number 98474-74-9.
- Available in small analytical pack sizes (e.g., 10 MG).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.