Learn More
Medchemexpress LLC Pseudoginsenoside RT5 | 98474-78-3 | 654.87 | 5 MG

Supplier: Medchemexpress LLC HYN05425MG
Pseudoginsenoside RT5 is a chemical compound isolated from Panax quinquefolium, intended for research use only. It appears as a white to off-white solid with a high purity of 99.32%.
- Isolated from Panax quinquefolium
- For research use only
- White to off-white solid appearance
- High purity of 99.32%
- Molecular weight of 654.87
- CAS Number: 98474-78-3
- Recommended storage for solid: 4°C, sealed, away from moisture and light
- Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month, sealed, away from moisture and light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.