Learn More
Medchemexpress LLC Pseudoisocyanine iodide | 977-96-8 | MFCD00011971 | 100.0% | 454.35 g/mol | C23H23IN2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10774010MM 1ML
Encompass_Preferred
Pseudoisocyanine iodide is a cyanine dye and biological transporter inhibitor (CAS 977-96-8) used in imaging, spectroscopy, and transporter inhibition assays. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid material for custom preparations.
  • Cyanine dye with transporter inhibition activity
  • Provided as a 10 mM solution in DMSO for ready-to-use stock solutions
  • Also available as solid forms for custom concentrations
  • High purity suitable for biochemical and cell-based assays
  • Molecular formula C23H23IN2, molecular weight 454.35 g/mol
Catalog No. 50-002-41186