Learn More
ATCC Pseudokirchneriella subcapitata
Supplier:  ATCC 22662
Encompass
Pseudokirchneriella subcapitata was isolated from the Nitelv River in Norway. This product has applications in algal growth-inhibition testing, toxicity testing, and algal growth potential testing.
Catalog No. 50-238-3925