Learn More
Medchemexpress LLC Pseudolaric Acid A | 82508-32-5 | 388.45 | 1 ML

Supplier: Medchemexpress LLC HYN067310MM/1ML
Pseudolaric Acid A is a diterpene acid isolated from Pseudolarix kaempferi. It exhibits antifungal, cytotoxic, and antifertile activities. This product is a 10 mM solution in 1 mL of DMSO and is intended for research use only.
- Purity: 99.90%
- Appearance: Solid
- Color: White to off-white
- Formula: C22H28O6
- Structure classification: Terpenoids, diterpenoids
- Initial source: Plants, Pinaceae, other families
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.