Learn More
Medchemexpress LLC Pseudolaric Acid B | 82508-31-4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN693910MG
Encompass_Preferred
Pseudolaric Acid B is an orally active diterpene acid. It exhibits anti-fungal, anti-fertility, anti-angiogenesis, and anticancer activities. It can induce apoptosis and autophagy in tumor cells. Additionally, it can inhibit the secretion of hepatitis B virus (HBV) and has an immunosuppressive effect, specifically inhibiting the proliferation of T lymphocytes and the production of IL-2.
  • Exhibits anti-fungal, anti-fertility, anti-angiogenesis, and anticancer activities
  • Induces apoptosis and autophagy in tumor cells
  • Inhibits secretion of hepatitis B virus (HBV)
  • Possesses immunosuppressive effect
  • Selectively inhibits proliferation of T lymphocytes and production of IL-2
Catalog No. 50-004-19847