Learn More
Medchemexpress LLC Pseudolaric Acid C | 82601-41-0 | 390.43 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN06725MG
Encompass_Preferred
Pseudolaric C is a diterpenoid that has been isolated from the root bark of the plant Pseudolarix amabilis. This compound exhibits antifungal activity.
  • Isolated from Pseudolarix amabilis.
  • Exhibits antifungal activity.
  • Available in solid form, white to off-white in appearance.
  • Soluble in DMSO at 100 mg/mL.
  • Has a purity of 99.74%.
  • Can be stored as a powder at -20°C for 3 years, or in solvent at -80°C for 6 months and -20°C for 1 month.
Catalog No. 50-003-88937