Learn More
ATCC Pseudomonas aeruginosa; GFP
Supplier:  ATCC 15692GFP
Encompass
This clone was derived from ATCC 15692 and contains a multicopy vector encoding the green fluorescent protein (Aequorea victoria) GFPmut3. This gene is expressed under the control of the Plac promoter (Escherichia coli). This construct has been designed for Gram-negative bacteria fluorescence labeling. Ampicillin resistance gene (bla) encoded on a plasmid. Confers resistance to 300 μg/mL ampicillin.
Catalog No. 50-238-4958
 Restricted Item