Learn More
ATCC Pseudomonas aeruginosa; Strain PAO1-LAC; genomic DNA
Supplier:  ATCC 502383911
Encompass
Genomic DNA from Pseudomonas aeruginosa strain PAO1-LAC (ATCC 47085). This product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3911
 Restricted Item