Learn More
ATCC Pseudomonas aeruginosa (Schroeter) Migula
Supplier:  ATCC 27853/DEL
Encompass
Pseudomonas aeruginosa strain Boston 41501 was isolated from blood culture. This whole-genome sequenced bacterial strain has applications in media and susceptibility testing, evaluation of Mueller-Hinton Agar, and as a quality control strain for Abbott, API, Autobac, BBL, bioMerieux Vitek, Micro-Media, MicroScan™, Roche Diagnostics, and Sensititre products.
Catalog No. 50-189-131FP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.