Learn More
ATCC Burkholderia diffusa
Supplier:  ATCC 700450
Encompass
Burkholderia diffusa strain JS872 is a bacterium that was isolated from soil and ground water. This strain is cited to degrade 2,4-dinitrotoluene.
Catalog No. 50-238-0034