Learn More
Medchemexpress LLC Pseudoprotodioscin | 102115-79-7 | 1031.18 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN06865MG
Encompass_Preferred
Pseudoprotodioscin is a furostanoside that inhibits SREBP1/2 and microRNA 33a/b levels, leading to a reduction in the gene expression responsible for the synthesis of cholesterol and triglycerides. It is intended for research use only.
  • Inhibits SREBP1/2 and microRNA 33a/b levels.
  • Reduces gene expression related to cholesterol and triglyceride synthesis.
  • Increases ABCA1 protein and mRNA levels.
  • Promotes ApoA-1-mediated cholesterol efflux.
  • Reduces HMGCR, FAS, and ACC mRNA levels.
  • Promotes low-density lipoprotein receptor by decreasing PCSK9 levels.
  • Purity: 99.17%.
Catalog No. 50-004-16557