Learn More
Cayman Chemical PseudoprotodIoscIn 50mg
SDP
Supplier:  Cayman Chemical 3321550 MG
Encompass_Preferred
A steroidal saponin; cytotoxic to A375, L929, and HeLa cancer cells (IC50s = 5.73, 5.09, and 3.32 µM, respectively); increases ERα nuclear accumulation in 3T3-L1 preadipocytes cultured in differentiation medium; inhibits LPS-induced increases in NF-κB activity in reporter assays using isolated mouse peritoneal macrophages and HUVECs; reduces atherosclerotic plaque area and inhibits increases in serum total cholesterol and triglyceride levels in ovariectomized ApoE-/- mice fed a high-cholesterol diet at 1, 2.5, and 5 mg/kg
Catalog No. 50-294-9116