Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Pseudorabies virus (Aujeszky strain) - VR-135, frozen preparation, BSL-2

Supplier:  ATCC VR135

Encompass

Pseudorabies virus (strain Aujeszky) is an ATCC-supplied laboratory strain intended for agricultural and virology research. The material is supplied frozen and requires Biosafety Level 2 containment; propagation is typically performed in permissive cell lines such as CRFK.

  • Biosafety level: BSL 2.
  • Product format: Frozen viral preparation.
  • Storage conditions: Store at -70°C or colder; stable in glycerol at 4°C.
  • Culture host: CRFK (feline kidney) cell line.
  • Incubation: 37°C for 2-4 days to observe cytopathic effects.
  • Observed effects: Cytopathic effect and syncytia formation in permissive cells.
  • Isolation source: Cow; geographic origin Hungary.
  • Verification: Whole-genome sequencing used for verification.

Catalog No. NC3477728


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.