Learn More
Medchemexpress LLC Psi-6206 | 863329-66-2 | C10H13FN2O5 | 1 ML

Supplier: Medchemexpress LLC HY1523610MM/1ML
PSI-6206 (RO 2433) is a deaminated derivative of PSI-6130, known as a potent and selective inhibitor of HCV NS5B polymerase. While it exhibits low potency in inhibiting HCV replicon with an EC90 of >100 μM, its 5'-triphosphate form (RO2433-TP) effectively inhibits RNA synthesis by both native HCV replicase and recombinant HCV polymerase NS5B. It has shown no activity or cytotoxicity in various anti-HCV assays.
- Deaminated derivative of PSI-6130
- Potent and selective inhibitor of HCV NS5B polymerase
- Exhibits low potency in inhibiting HCV replicon (EC90 >100 μM)
- No activity or cytotoxicity observed in anti-HCV assays
- 5'-triphosphate form inhibits RNA synthesis by native HCV replicase and recombinant HCV polymerase NS5B
- Purity: 99.89%
- Molecular weight: 260.22
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.