Learn More
Medchemexpress LLC HY-15526 1mg Medchemexpress, PSI-697 CAS:851546-61-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY155261MG
Encompass_Preferred
Medchemexpress, HY-15526 1mg PSI-697 CAS:851546-61-7 PSI-697 is an oral P-selectin inhibitor with an IC50 of 125 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1731