Learn More
Abcam Psicofuranine, Adenosine analog, 5MG
Supplier:  Abcam AB1443365MG
Encompass_Preferred
MW 297.27 Da, Purity >98%. Adenosine analog. Antineoplastic and antibiotic agent. Blocks guanosine monophosphate synthesis. Inhibits xanthosine monophosphate. Reduces phosphate utilization in tumors. Shows antitumor effects and antibacterial activity in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5869