Learn More
AdipoGen Psicofuranine Angustmycin C
SDP
Supplier:  AdipoGen BVT0284M005
Encompass_Preferred
Chemical. CAS 1874-54-0. Formula C11H15N5O5. MW 297.3. Isolated from Streptomyces sp. S 2113. Nucleoside antibiotic. Antitumor compound. Xanthosine monophosphate XMP aminase inhibitor. Antimetabolite of the purine biosynthesis.
Catalog No. 50-256-2016