Learn More
Medchemexpress LLC Psma-11 (HBED-CC-PSMA) | 1366302-52-4 | 99.7% | 946.99 | C44H62N6O17 | 100MG
SDP
Supplier:  Medchemexpress LLC HY125399100MG
Encompass_Preferred
PSMA-11 is a small-molecule ligand that binds prostate-specific membrane antigen (PSMA) and is used as a precursor for synthesizing gallium-68 labeled PET tracers for prostate cancer imaging. It is supplied for research use and is characterized by analytical data to support identity and purity.
  • Suitable precursor for radiolabeling with gallium-68.
  • High purity (>99.7%) for research applications.
  • Characterized by HNMR and LCMS data for identity confirmation.
  • Recommended storage: solid at -20°C, protect from light.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-35700