Learn More
Medchemexpress LLC PSMA-Val-Cit-PAB-MMAE | 2748039-79-2 | 99.5% | 2267.10 | C114H165ClN20O26 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1418601MG
Encompass_Preferred
PSMA-Val-Cit-PAB-MMAE is a small-molecule PSMA-targeted conjugate that uses a valine-citrulline (Val-Cit) peptide linker and a para-aminobenzyl (PAB) spacer to release the cytotoxic payload monomethyl auristatin E (MMAE) for research into PSMA-targeted chemotherapy of prostate cancer.
  • Small-molecule PSMA-targeted conjugate based on monomethyl auristatin E.
  • High purity (99.52%) and defined molecular formula and weight.
  • White to off-white solid appearance for ease of handling.
  • Storage: powder stable at -20°C for 3 years; in solvent stability at -80°C for 6 months.
  • Intended for research use only.
Catalog No. 50-002-03772